Assay Details
Target Gene Details
Entrez Gene ID: | 56455 |
Gene Name: | dynein light chain LC8-type 1 |
Gene Aliases: |
Dlc8, Dnclc1, Pin |
Location: |
Chr.5:115297110-115300990 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dynll1 | NM_019682.4 | NP_062656.3 | ||
AF020185.1 | AAD01643.1 | |||
AK002522.1 | BAB22160.1 | |||
AK010614.1 | BAB27063.1 | |||
AK010685.1 | BAB27117.1 | |||
AK013721.1 | BAB28970.1 | |||
AK013725.1 | BAB28973.1 | |||
AK082923.1 | BAC38691.1 | |||
AK147977.1 | BAE28262.1 | |||
BC008106.1 | AAH08106.1 | |||
BC034258.1 | AAH34258.1 | |||
CA465333.1 | ||||
CN832868.1 |
More Information
Set Membership: |
Intragenic Non-exonic |