Assay Details
Target Gene Details
Entrez Gene ID: | 67980 |
Gene Name: | glucosamine-6-phosphate deaminase 2 |
Gene Aliases: |
4921523I18Rik, 4933412A11Rik, BB189630, GNPDA, Gnp2, Sb52, mKIAA4008 |
Location: |
Chr.5:69574998-69592332 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gnpda2 | NM_001038015.1 | 5 | 659 | NP_001033104.1 |
AK014950.1 | 5 | 669 | ||
AK015527.1 | 5 | 669 | BAB29883.1 | |
AK017588.1 | 5 | 668 | BAB30824.1 | |
AK051596.1 | 5 | 668 | ||
AK141408.1 | 5 | 671 | BAE24675.1 | |
AK166601.1 | 5 | 661 | BAE38886.1 | |
AK168516.1 | 5 | 678 | BAE40396.1 | |
AK171299.1 | 5 | 654 | BAE42378.1 | |
AK220275.1 | 5 | 3221 | BAD90200.1 | |
BC004084.1 | 5 | 659 | AAH04084.1 |
More Information
Set Membership: |
Intragenic Exonic |