Assay Details
Target Gene Details
Entrez Gene ID: | 114674 |
Gene Name: | GTF2I repeat domain containing 2 |
Gene Aliases: |
1700012P16Rik |
Location: |
Chr.5:134183215-134218146 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gtf2ird2 | NM_053266.1 | NP_444496.1 | ||
AK081801.1 | ||||
AY014963.1 | AAG41674.1 | |||
AY116023.1 | AAM48282.1 | |||
BC132157.1 | ||||
BC132159.1 |
More Information
Set Membership: |
Intragenic Non-exonic |