Assay Details
Target Gene Details
Entrez Gene ID: | 114674 |
Gene Name: | GTF2I repeat domain containing 2 |
Gene Aliases: |
1700012P16Rik |
Location: |
Chr.5:134183215-134218146 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gtf2ird2 | NM_053266.1 | 3 | 389 | NP_444496.1 |
AK081801.1 | 3 | 398 | ||
AY014963.1 | 3 | 389 | AAG41674.1 | |
AY116023.1 | 3 | 400 | AAM48282.1 | |
BC132157.1 | 3 | 297 | ||
BC132159.1 | 3 | 297 |
More Information
Set Membership: |
Intragenic Exonic |