Assay Details
Target Gene Details
Entrez Gene ID: | 15234 |
Gene Name: | hepatocyte growth factor |
Gene Aliases: |
C230052L06Rik, HGF/SF, NK1, NK2, SF, SF/HGF |
Location: |
Chr.5:16553335-16622408 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hgf | NM_001289458.1 | 2 | 238 | NP_001276387.1 |
NM_001289459.1 | 2 | 227 | NP_001276388.1 | |
NM_001289460.1 | 2 | 238 | NP_001276389.1 | |
NM_001289461.1 | 1 | 60 | NP_001276390.1 | |
NM_010427.5 | 2 | 210 | NP_034557.3 | |
AF042856.1 | AAC40051.1 | |||
AK042121.1 | 2 | 156 | BAC31175.1 | |
AK052289.1 | 2 | 239 | ||
AK082461.1 | 1 | 210 | BAC38497.1 | |
AK142159.1 | 2 | 156 | ||
BC119228.1 | 2 | 97 | ||
D10212.1 | BAA01064.1 | |||
D10213.1 | BAA01065.1 | |||
S71816.1 | AAB31855.1 | |||
X72307.1 | 1 | 94 | CAA51054.1 | |
X84046.1 | 1 | 73 | CAA58865.1 |
More Information
Set Membership: |
Intragenic Exonic |