Assay Details
Target Gene Details
Entrez Gene ID: | 27979 |
Gene Name: | eukaryotic translation initiation factor 3, subunit B |
Gene Aliases: |
AL033316, AL033334, AL033369, AW208965, D5Wsu45e, EIF3-ETA, EIF3-P110, EIF3-P116, Eif3s9, PRT1 |
Location: |
Chr.5:140419305-140443358 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif3b | NM_133916.2 | 14 | 1976 | NP_598677.1 |
AK033722.1 | 14 | 1976 | BAC28445.1 | |
AK089978.1 | 14 | 1965 | ||
AK167618.1 | 14 | 1963 | BAE39671.1 | |
AK170938.1 | 14 | 1976 | BAE42128.1 | |
AK205605.1 | ||||
BC007175.1 | 13 | 1435 | AAH07175.1 | |
BC023767.1 | 14 | 1947 | AAH23767.1 | |
BC031704.1 | 14 | 1953 | AAH31704.1 | |
BC051065.1 | 14 | 1936 | AAH51065.1 |
More Information
Set Membership: |
Intragenic Exonic |