Assay Details
Target Gene Details
Entrez Gene ID: | 94244 |
Gene Name: | FK506 binding protein 6 |
Gene Aliases: |
1700008G22Rik, 36kDa, AU017274, D5Ertd724e, FKBP-36, FKBP-6 |
Location: |
Chr.5:135291704-135351807 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fkbp6 | NM_001277891.1 | NP_001264820.1 | ||
NM_001277892.1 | NP_001264821.1 | |||
NM_001277893.1 | NP_001264822.1 | |||
NM_033571.3 | NP_291049.1 | |||
AF367709.1 | AAK53411.1 | |||
AF367710.1 | AAK53412.1 | |||
AF367711.1 | AAK53413.1 | |||
AK090036.1 | BAC41058.1 | |||
AY029192.1 | AAK39645.1 | |||
BC139092.1 | ||||
BC139097.1 |
More Information
Set Membership: |
Intragenic Non-exonic |