Assay Details
Target Gene Details
Entrez Gene ID: | 242915 |
Gene Name: | GRB2 associated, regulator of MAPK1-like |
Gene Aliases: |
Fam59b, Gm444 |
Location: |
Chr.5:30105196-30118380 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gareml | NM_001167879.1 | 3 | 415 | NP_001161351.1 |
BC060268.1 | 2 | 199 | AAH60268.1 | |
BC113782.1 | 2 | 845 | AAI13783.1 | |
BC113783.1 | 2 | 846 | ||
CX204487.1 |