Assay Details
Target Gene Details
Entrez Gene ID: | 56378 |
Gene Name: | actin related protein 2/3 complex, subunit 3 |
Gene Aliases: |
1110006A04Rik, 21kDa, AA408672, AI788639, p21-Ar, p21Arc |
Location: |
Chr.5:122391928-122406178 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arpc3 | NM_019824.3 | NP_062798.1 | ||
AB038243.1 | BAA90788.1 | |||
AK003485.1 | BAB22813.1 | |||
AK005437.1 | BAB24031.1 | |||
AK171651.1 | BAE42588.1 | |||
BC013618.1 | AAH13618.1 | |||
BC054440.1 | AAH54440.1 |
More Information
Set Membership: |
Intragenic Non-exonic |