Assay Details
Target Gene Details
Entrez Gene ID: | 330122 |
Gene Name: | chemokine (C-X-C motif) ligand 3 |
Gene Aliases: |
Dcip1, Gm1960 |
Location: |
Chr.5:90786101-90788093 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cxcl3 | NM_203320.3 | 1 | 34 | NP_976065.1 |
AK144158.1 | 1 | 37 | BAE25738.1 | |
AK150314.1 | BAE29460.1 | |||
AK154334.1 | BAE32520.1 | |||
AY311403.1 | AAQ86936.1 | |||
BC117014.1 | ||||
BC117016.1 |
More Information
Set Membership: |
Intragenic Exonic |