Assay Details
Target Gene Details
Entrez Gene ID: | 381680 |
Gene Name: | neurexophilin and PC-esterase domain family, member 5 |
Gene Aliases: |
- |
Location: |
Chr.5:138207499-138251876 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nxpe5 | NM_001013773.3 | NP_001013795.3 | ||
AK156114.1 |