Assay Details
Target Gene Details
Entrez Gene ID: | 12571 |
Gene Name: | cyclin-dependent kinase 6 |
Gene Aliases: |
5830411I20, AI504062, Crk2 |
Location: |
Chr.5:3343891-3531008 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdk6 | NM_009873.2 | NP_034003.1 | ||
AF132482.1 | AAD43503.1 | |||
AF132483.1 | AAD43504.1 | |||
AK078973.1 | BAC37489.1 | |||
AK154197.1 | BAE32432.1 | |||
BC119360.1 | ||||
BC120595.1 |
More Information
Set Membership: |
Intragenic Non-exonic |