Assay Details
Target Gene Details
Entrez Gene ID: | 231162 |
Gene Name: | cytokine-like 1 |
Gene Aliases: |
4930443F05Rik, 4Cytl1, C17, Cyt1, Gm147 |
Location: |
Chr.5:37735519-37739820 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cytl1 | NM_001081106.1 | 3 | 297 | NP_001074575.1 |
AK076770.1 | 2 | 189 | ||
BC063103.1 | 3 | 257 | AAH63103.1 | |
BC116224.1 | 3 | 271 | ||
BC116225.1 | 3 | 271 | ||
EF108311.1 | 3 | 268 |
More Information
Set Membership: |
Intragenic Exonic |