Assay Details
Target Gene Details
Entrez Gene ID: | 74364 |
Gene Name: | RIKEN cDNA 4931431C16 gene |
Gene Aliases: |
- |
Location: |
Chr.5:35581227-35588763 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4931431C16Rik | NR_045807.1 | |||
AK016496.1 | BAC25484.1 | |||
AK030185.1 | BAC26829.1 |
Target Gene Details
Entrez Gene ID: | 80911 |
Gene Name: | acyl-Coenzyme A oxidase 3, pristanoyl |
Gene Aliases: |
BI685180, EST-s59, PCOX |
Location: |
Chr.5:35582967-35614210 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Acox3 | NM_030721.2 | 3 | 254 | NP_109646.2 |
AJ278430.1 | 3 | 287 | CAC20692.1 | |
AK028818.1 | 3 | 675 | BAC26136.1 | |
AK053960.1 | 3 | 251 | ||
AK128922.1 | 3 | 214 | ||
AK154112.1 | 2 | 345 | ||
AK170083.1 | ||||
BC044725.2 | 3 | 274 | AAH44725.1 | |
BC055019.1 | 3 | 254 | AAH55019.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |