Assay Details
Target Gene Details
Entrez Gene ID: | 74364 |
Gene Name: | RIKEN cDNA 4931431C16 gene |
Gene Aliases: |
- |
Location: |
Chr.5:35581227-35588763 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4931431C16Rik | NR_045807.1 | |||
AK016496.1 | BAC25484.1 | |||
AK030185.1 | BAC26829.1 |
Target Gene Details
Entrez Gene ID: | 80911 |
Gene Name: | acyl-Coenzyme A oxidase 3, pristanoyl |
Gene Aliases: |
BI685180, EST-s59, PCOX |
Location: |
Chr.5:35582967-35614210 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Acox3 | NM_030721.2 | NP_109646.2 | ||
AJ278430.1 | CAC20692.1 | |||
AK028818.1 | BAC26136.1 | |||
AK053960.1 | ||||
AK080942.1 | 2 | 179 | ||
AK128922.1 | ||||
AK154112.1 | ||||
AK170083.1 | ||||
BC044725.2 | AAH44725.1 | |||
BC055019.1 | AAH55019.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |