Assay Details
Target Gene Details
Entrez Gene ID: | 12962 |
Gene Name: | crystallin, beta B3 |
Gene Aliases: |
AI852419 |
Location: |
Chr.5:113075839-113082233 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Crybb3 | NM_001159650.1 | 6 | 691 | NP_001153122.1 |
NM_021352.3 | 6 | 761 | NP_067327.1 | |
AJ272229.2 | 5 | 745 | CAB75587.1 | |
BC031442.1 | 5 | 752 | AAH31442.1 | |
BC058522.1 | 5 | 670 | AAH58522.1 |