Assay Details
Target Gene Details
Entrez Gene ID: | 56221 |
Gene Name: | chemokine (C-C motif) ligand 24 |
Gene Aliases: |
CKb-6, MPIF-2, Scya24 |
Location: |
Chr.5:135569937-135573122 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccl24 | NM_019577.4 | NP_062523.1 | ||
AF244367.1 | AAF61736.1 | |||
AF281075.1 | AAG32664.1 | |||
AK009718.1 | BAE20423.1 | |||
AK157068.1 | BAE33951.1 | |||
BC065389.1 | AAH65389.1 |
More Information
Set Membership: |
Intragenic Non-exonic |