Assay Details
Target Gene Details
Entrez Gene ID: | 66096 |
Gene Name: | late endosomal/lysosomal adaptor, MAPK and MTOR activator 4 |
Gene Aliases: |
0910001L09Rik, AV006840 |
Location: |
Chr.5:138255482-138259395 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lamtor4 | NM_001081108.2 | NP_001074577.1 | ||
AK002488.1 | BAC24993.1 | |||
AK003086.1 | BAC25020.1 | |||
AK003250.1 | ||||
AK010598.1 | BAC25306.1 | |||
AK146994.1 | ||||
BC144958.1 |