Assay Details
Target Gene Details
Entrez Gene ID: | 230908 |
Gene Name: | TAR DNA binding protein |
Gene Aliases: |
1190002A23Rik, C85084, TDP-43, Tdp43 |
Location: |
Chr.4:148612382-148626996 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tardbp | NM_001003898.3 | NP_001003898.1 | ||
NM_001003899.2 | NP_001003899.1 | |||
NM_001008545.2 | NP_001008545.1 | |||
NM_001008546.2 | NP_001008546.1 | |||
NM_145556.4 | NP_663531.1 | |||
NR_027864.1 | ||||
AK032199.1 | BAC27753.1 | |||
AK045495.1 | BAC32395.1 | |||
AK076208.1 | ||||
AK083312.1 | BAC38861.1 | |||
AK133207.1 | BAE21557.1 | |||
AK146002.1 | BAE26823.1 | |||
AK146997.1 | ||||
AK153803.1 | BAE32189.1 | |||
AY319513.1 | AAP92688.1 | |||
AY319514.1 | AAP92689.1 | |||
AY319515.1 | AAP92690.1 | |||
AY319516.1 | AAP92691.1 | |||
BC012873.1 | AAH12873.1 | |||
BC025544.1 | AAH25544.1 | |||
BC027105.1 | AAH27105.1 | |||
BC027772.1 | AAH27772.1 | |||
BC029728.1 | ||||
BC031126.1 | AAH31126.1 | |||
BC033475.1 | AAH33475.1 | |||
GQ428775.1 | ||||
GQ428776.1 | ||||
GQ428777.1 | ||||
GU250735.1 |