Assay Details
Target Gene Details
Entrez Gene ID: | 100434 |
Gene Name: | solute carrier family 44, member 1 |
Gene Aliases: |
2210409B22Rik, 4833416H08Rik, AW547365, CHTL1, CTL1, Cdw92 |
Location: |
Chr.4:53440413-53622478 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc44a1 | NM_001159633.1 | 8 | 1094 | NP_001153105.1 |
NM_133891.3 | 8 | 1094 | NP_598652.3 | |
AK077951.1 | 8 | 1097 | ||
AK200057.1 | 3 | 252 | ||
AY249865.1 | 8 | 850 | AAP81042.1 | |
AY249866.1 | 8 | 823 | AAP81043.1 | |
BC113167.1 | 8 | 850 | ||
BC113169.1 | 8 | 850 | ||
CF532681.1 |
More Information
Set Membership: |
Intragenic Exonic |