Assay Details
Target Gene Details
Entrez Gene ID: | 20731 |
Gene Name: | serine peptidase inhibitor, Kazal type 4 |
Gene Aliases: |
MPGC60 |
Location: |
Chr.4:40920056-40931395 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Spink4 | NM_011463.2 | NP_035593.2 | ||
BC117048.1 | ||||
BC117050.1 | ||||
CF381698.1 |
More Information
Set Membership: |
Intragenic Non-exonic |