Assay Details
Target Gene Details
Entrez Gene ID: | 230484 |
Gene Name: | ubiquitin specific peptidase 1 |
Gene Aliases: |
- |
Location: |
Chr.4:98923622-98935543 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Usp1 | NM_001301414.1 | 2 | 520 | NP_001288343.1 |
NM_146144.4 | 2 | 501 | NP_666256.2 | |
AK080882.1 | 2 | 502 | BAC38059.1 | |
BC018179.1 | 2 | 381 | AAH18179.1 | |
BC020007.1 | 2 | 353 | AAH20007.1 | |
BY713495.1 | 2 | 389 |
More Information
Set Membership: |
Intragenic Exonic |