Assay Details
Target Gene Details
Entrez Gene ID: | 64817 |
Gene Name: | sushi, von Willebrand factor type A, EGF and pentraxin domain containing 1 |
Gene Aliases: |
1110021D17Rik, 4833413O10Rik, AA387071, D430029O09Rik, Polydom |
Location: |
Chr.4:58042796-58206596 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 34 - Exon 34 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Svep1 | NM_022814.2 | NP_073725.2 | ||
AF206329.1 | AAG32160.1 |