Assay Details
Target Gene Details
Entrez Gene ID: | 21812 |
Gene Name: | transforming growth factor, beta receptor I |
Gene Aliases: |
ALK5, AU017191, Alk-5, TbetaR-I, TbetaRI |
Location: |
Chr.4:47353113-47414926 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tgfbr1 | NM_009370.2 | 3 | 411 | NP_033396.1 |
AK015337.1 | 2 | 347 | BAB29802.1 | |
AK154381.1 | 2 | 174 | BAE32548.1 | |
BC063260.1 | 3 | 411 | AAH63260.1 | |
CT010393.1 | 3 | 404 | CAJ18600.1 | |
D25540.1 | 3 | 397 | BAA05023.1 | |
D28526.1 | 3 | 404 | BAA05878.1 |
More Information
Set Membership: |
Intragenic Exonic |