Assay Details
Target Gene Details
Entrez Gene ID: | 70977 |
Gene Name: | calcium binding protein, spermatid specific 1 |
Gene Aliases: |
4931407G18Rik, AI428159, Clph |
Location: |
Chr.5:87979451-87981541 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cabs1 | NM_027631.2 | NP_081907.1 | ||
AK016441.1 | BAB30237.1 | |||
AK076636.1 | BAC36426.1 | |||
BC049750.1 | AAH49750.1 |
Target Gene Details
Entrez Gene ID: | 20599 |
Gene Name: | submaxillary gland androgen regulated protein 3A |
Gene Aliases: |
AA589601, MSG1, MSG3, Smr1, Smr3 |
Location: |
Chr.5:87969504-88008534 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smr3a |