Assay Details
Target Gene Details
Entrez Gene ID: | 100340 |
Gene Name: | sphingomyelin phosphodiesterase, acid-like 3B |
Gene Aliases: |
1110054A24Rik, AU045240, Asml3b |
Location: |
Chr.4:132732966-132757171 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smpdl3b | NM_133888.2 | 4 | 570 | NP_598649.1 |
AK136602.1 | 3 | 232 | ||
AK154599.1 | 4 | 596 | BAE32702.1 | |
AK166253.1 | 4 | 546 | BAE38663.1 | |
BC009087.1 | 4 | 570 | AAH09087.1 |
More Information
Set Membership: |
Intragenic Exonic |