Assay Details
Target Gene Details
Entrez Gene ID: | 654318 |
Gene Name: | RIKEN cDNA C530005A16 gene |
Gene Aliases: |
D930044c15rik |
Location: |
Chr.4:116589733-116597630 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C530005A16Rik | NR_029450.1 | 1 | 741 | |
AK035052.1 | 1 | 746 | ||
AK039789.1 | 1 | 744 | ||
AK086654.1 | 1 | 742 | BAC39711.1 | |
AK140096.1 | 1 | 722 | ||
AK164083.1 | 1 | 706 |
Target Gene Details
Entrez Gene ID: | 622665 |
Gene Name: | coiled-coil domain containing 17 |
Gene Aliases: |
- |
Location: |
Chr.4:116596730-116600266 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccdc17 | NM_001037916.3 | 1 | 161 | NP_001033005.1 |
AK029257.1 | 1 | 163 | BAC26359.1 | |
AK076542.1 | 1 | 238 | ||
BC150874.1 | 1 | 146 | ||
BC172023.1 | 1 | 146 |