Assay Details
Target Gene Details
Entrez Gene ID: | 76376 |
Gene Name: | solute carrier family 24 (sodium/potassium/calcium exchanger), member 2 |
Gene Aliases: |
2810021B17Rik, 6330417K15Rik, AI847460, Nckx2 |
Location: |
Chr.4:86983124-87230554 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc24a2 | NM_001110240.1 | 2 | 1174 | NP_001103710.1 |
NM_172426.2 | NP_766014.1 | |||
AK034424.1 | 3 | 1283 | BAC28705.1 | |
AK038634.1 | ||||
AK038800.1 | 3 | 1327 | BAC30134.1 | |
AK083127.1 | BAC38773.1 | |||
BC115867.1 | 2 | 1176 | ||
BC116637.1 |
More Information
Set Membership: |
Intragenic Exonic |