Assay Details
Target Gene Details
Entrez Gene ID: | 22178 |
Gene Name: | tyrosinase-related protein 1 |
Gene Aliases: |
Oca3, TRP-1, TRP1, Tyrp, b, brown, isa |
Location: |
Chr.4:80834208-80851736 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tyrp1 | NM_001282014.1 | 5 | 1269 | NP_001268943.1 |
NM_001282015.1 | 5 | 1263 | NP_001268944.1 | |
NM_031202.3 | 5 | 1284 | NP_112479.1 | |
AK148332.1 | 5 | 1269 | BAE28487.1 | |
AK148370.1 | 5 | 1175 | BAE28511.1 | |
AK148441.1 | 5 | 1175 | BAE28554.1 | |
AK180622.1 | 2 | 186 | ||
AK190391.1 | 2 | 186 | ||
AK190783.1 | 2 | 186 | ||
AK191129.1 | 2 | 186 | ||
AK191532.1 | 2 | 186 | ||
AK191540.1 | 2 | 186 | ||
AK193049.1 | 2 | 186 | ||
AK193925.1 | 2 | 186 | ||
AK194181.1 | 2 | 186 | ||
AK194480.1 | 2 | 186 | ||
AK194755.1 | 2 | 160 | ||
AK196212.1 | 2 | 186 | ||
AK196465.1 | 2 | 186 | ||
AK197240.1 | 2 | 186 | ||
AK197400.1 | 2 | 186 | ||
AK197957.1 | 2 | 186 | ||
AK199510.1 | 2 | 186 | ||
AK199962.1 | 2 | 186 | ||
AK200845.1 | 2 | 186 | ||
AK201045.1 | 2 | 186 | ||
AK201297.1 | 2 | 186 | ||
AK202385.1 | 2 | 186 | ||
AK203202.1 | 2 | 186 | ||
AK203420.1 | 2 | 186 | ||
AK204585.1 | 2 | 187 | ||
AK204597.1 | 2 | 186 | ||
AK204783.1 | 2 | 186 | ||
AK205230.1 | 2 | 186 | ||
AK209301.1 | 2 | 185 | ||
AK211654.1 | 2 | 186 | ||
AK214877.1 | 2 | 186 | ||
AK219979.1 | 2 | 185 | ||
BC076598.1 | 5 | 1267 | AAH76598.1 | |
X03687.1 | 5 | 1169 | CAA27323.1 |
More Information
Set Membership: |
Intragenic Exonic |