Assay Details
Target Gene Details
Entrez Gene ID: | 106021 |
Gene Name: | topoisomerase I binding, arginine/serine-rich |
Gene Aliases: |
AW105885, LUN, TP53BPL, p53BP3/LUN |
Location: |
Chr.4:40259606-40269841 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Topors | NM_134097.3 | NP_598858.2 | ||
AB072395.1 | BAB69457.1 | |||
AB104865.1 | BAC65157.1 | |||
AK044564.1 | BAC31981.2 | |||
AK134075.1 | BAE22003.1 | |||
AK140250.1 | BAE24298.1 | |||
AK143025.1 | BAE25253.1 | |||
AK153743.1 | BAE32164.1 | |||
BC019421.1 | ||||
BC037141.1 | AAH37141.1 | |||
BC040797.1 | AAH40797.1 |
More Information
Set Membership: |
Intragenic Non-exonic |