Assay Details
Target Gene Details
Entrez Gene ID: | 230157 |
Gene Name: | transmembrane protein with EGF-like and two follistatin-like domains 1 |
Gene Aliases: |
A830033E11, M7365, Tr1 |
Location: |
Chr.4:48585114-48663131 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmeff1 | NM_021436.2 | 9 | 1270 | NP_067411.1 |
AJ400622.1 | 9 | 939 | CAB90827.1 | |
AK043792.1 | 7 | 646 | BAC31655.1 | |
AK079633.1 | 7 | 620 | BAC37709.1 | |
BC057598.1 | 9 | 1270 | AAH57598.1 |
More Information
Set Membership: |
Intragenic Exonic |