Assay Details
Target Gene Details
Entrez Gene ID: | 67103 |
Gene Name: | prostaglandin reductase 1 |
Gene Aliases: |
2510002C21Rik, Ltb4dh, Zadh3 |
Location: |
Chr.4:58965590-58987131 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptgr1 | NM_025968.3 | NP_080244.1 | ||
AK010888.1 | BAB27248.1 | |||
AK011962.1 | BAB27941.1 | |||
AK035425.1 | BAC29060.1 | |||
AK134440.1 | BAE22145.1 | |||
AK166835.1 | BAE39057.1 | |||
BC014865.1 | AAH14865.1 | |||
CT010267.1 | CAJ18475.1 |
More Information
Set Membership: |
Intragenic Non-exonic |