Assay Details
Target Gene Details
Entrez Gene ID: | 26970 |
Gene Name: | phospholipase A2, group IIE |
Gene Aliases: |
- |
Location: |
Chr.4:138877911-138882817 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pla2g2e | NM_012044.2 | 3 | 266 | NP_036174.1 |
AF112984.1 | 3 | 266 | AAF22290.1 | |
AF166098.1 | 3 | 262 | AAF04499.1 | |
BC027524.1 | 2 | 155 | AAH27524.1 |
More Information
Set Membership: |
Intragenic Exonic |