Assay Details
Target Gene Details
Entrez Gene ID: | 12695 |
Gene Name: | InaD-like (Drosophila) |
Gene Aliases: |
Cipp, Patj |
Location: |
Chr.4:98395534-98719603 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Inadl | NM_001005784.1 | 18 | 2313 | NP_001005784.1 |
NM_172696.2 | 18 | 2313 | NP_766284.2 | |
AK050545.1 | ||||
AK077268.1 | 5 | 595 | BAC36720.1 | |
AK147866.1 | 18 | 2262 | BAE28189.1 | |
AK153597.1 | 7 | 1168 | BAE32113.1 | |
AK156166.1 | 7 | 781 | BAE33609.1 | |
BB617492.1 | 5 | 592 | ||
BC062194.1 | 6 | 615 | AAH62194.1 | |
BC082787.1 | 7 | 706 | AAH82787.1 | |
BC141404.1 | 18 | 2289 | ||
CD547724.1 | 2 | 185 |