Assay Details
Target Gene Details
Entrez Gene ID: | 67096 |
Gene Name: | methylmalonic aciduria cblC type, with homocystinuria |
Gene Aliases: |
1810037K07Rik |
Location: |
Chr.4:116702434-116708385 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mmachc | NM_025962.3 | NP_080238.2 | ||
AK007725.1 | BAB25214.1 | |||
AK012761.1 | BAB28451.1 | |||
AK084194.1 | BAC39135.1 | |||
AK213975.1 | ||||
BB614283.1 | ||||
BC054756.1 | AAH54756.1 |