Assay Details
Target Gene Details
Entrez Gene ID: | 12795 |
Gene Name: | polo-like kinase 3 |
Gene Aliases: |
Cnk, Fnk, PRK |
Location: |
Chr.4:117128655-117133952 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plk3 | NM_013807.2 | NP_038835.2 | ||
AF136586.1 | AAF08369.1 | |||
AK156754.1 | BAE33837.1 | |||
BC063051.1 | AAH63051.1 | |||
U21392.1 | AAC52191.1 |
More Information
Set Membership: |
Intragenic Non-exonic |