Assay Details
Target Gene Details
Entrez Gene ID: | 231002 |
Gene Name: | pleckstrin homology domain containing, family N member 1 |
Gene Aliases: |
BC025458 |
Location: |
Chr.4:156221456-156228542 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plekhn1 | NM_001008233.3 | NP_001008234.2 | ||
AK048084.1 | BAC33236.1 | |||
BC025458.1 | AAH25458.1 |