Assay Details
Target Gene Details
Entrez Gene ID: | 230899 |
Gene Name: | natriuretic peptide type A |
Gene Aliases: |
ANP, Anf, Pnd |
Location: |
Chr.4:148000746-148002067 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nppa | NM_008725.2 | 1 | 61 | NP_032751.1 |
AK147180.1 | 1 | 65 | BAE27742.1 | |
BC089615.1 | 1 | 61 | AAH89615.1 | |
KC526925.1 | ||||
KC526926.1 | ||||
KC526927.1 |