Assay Details
Target Gene Details
Entrez Gene ID: | 50927 |
Gene Name: | nuclear autoantigenic sperm protein (histone-binding) |
Gene Aliases: |
5033430J04Rik, AI131596, AI317140, D4Ertd767e, Epcs32, Nasp-T |
Location: |
Chr.4:116601052-116627951 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nasp | NM_001081475.1 | NP_001074944.1 | ||
NM_001284229.1 | NP_001271158.1 | |||
NM_016777.3 | NP_058057.3 | |||
AF034610.2 | AAB87567.2 | |||
AF095722.1 | AAC64195.1 | |||
AK083333.1 | BAC38871.1 | |||
BC004693.1 | AAH04693.1 |
More Information
Set Membership: |
Intragenic Non-exonic |