Assay Details
Target Gene Details
Entrez Gene ID: | 19266 |
Gene Name: | protein tyrosine phosphatase, receptor type, D |
Gene Aliases: |
1110002J03Rik, 3000002J10Rik, B230219D21Rik, R-PTP-delta |
Location: |
Chr.4:75941237-78211895 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 35 - Exon 36 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprd | NM_011211.3 | NP_035341.2 | ||
AF326559.1 | AAL37405.1 | |||
AF326560.1 | AAL37406.1 | |||
AK043201.1 | BAC31488.1 | |||
AK051809.1 | BAE43348.1 | |||
BC025145.1 | AAH25145.1 | |||
BC141409.1 | ||||
D13903.1 | BAA03003.1 |
More Information
Set Membership: |
Intragenic Non-exonic |