Assay Details
Target Gene Details
Entrez Gene ID: | 67182 |
Gene Name: | PDZK1 interacting protein 1 |
Gene Aliases: |
0610007F13Rik, 2700030M23Rik, AU046042, Map17 |
Location: |
Chr.4:115088708-115093894 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pdzk1ip1 | NM_001164557.1 | 2 | 558 | NP_001158029.1 |
NM_001164558.1 | 3 | 255 | NP_001158030.1 | |
NM_026018.3 | 3 | 326 | NP_080294.1 | |
AB009296.1 | 2 | 136 | ||
AK002288.1 | 1 | 263 | ||
AK007936.1 | 2 | 544 | BAB25361.1 | |
AK008253.1 | 3 | 312 | BAB25559.1 | |
AK012305.1 | 3 | 273 | BAB28152.1 | |
BC013542.1 | 2 | 163 | AAH13542.1 | |
DV057288.1 | 3 | 272 | ||
DV058382.1 | 2 | 247 |