Assay Details
Target Gene Details
Entrez Gene ID: | 230145 |
Gene Name: | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 12 |
Gene Aliases: |
9130206E10, A630062B03Rik |
Location: |
Chr.4:47091893-47123042 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Galnt12 | NM_172693.3 | NP_766281.1 | ||
AB175680.1 | BAD52068.1 | |||
AK033638.1 | BAC28401.1 | |||
AK042133.1 | BAC31179.1 | |||
AK043977.1 | ||||
AK082014.1 | ||||
AK156751.1 | BAE33836.1 | |||
BC056425.1 | AAH56425.1 |
More Information
Set Membership: |
Intragenic Non-exonic |