Assay Details
Target Gene Details
Entrez Gene ID: | 332942 |
Gene Name: | predicted gene 853 |
Gene Aliases: |
OTTMUSG00000009560 |
Location: |
Chr.4:130209023-130222427 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm853 | NM_001034872.2 | NP_001030044.1 | ||
AK143920.1 | BAE25605.1 | |||
BC147394.1 | ||||
BC147395.1 | ||||
BC172060.1 |
More Information
Set Membership: |
Intragenic Non-exonic |