Assay Details
Target Gene Details
Entrez Gene ID: | 332942 |
Gene Name: | predicted gene 853 |
Gene Aliases: |
OTTMUSG00000009560 |
Location: |
Chr.4:130209023-130222427 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm853 | NM_001034872.2 | 2 | 218 | NP_001030044.1 |
AK143920.1 | 2 | 219 | BAE25605.1 | |
BC147394.1 | 1 | 83 | ||
BC147395.1 | 1 | 83 | ||
BC172060.1 | 1 | 83 |
More Information
Set Membership: |
Intragenic Exonic |