Assay Details
Target Gene Details
Entrez Gene ID: | 12804 |
Gene Name: | ciliary neurotrophic factor receptor |
Gene Aliases: |
Cntfralpha |
Location: |
Chr.4:41657498-41697091 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cntfr | NM_001136056.2 | NP_001129528.1 | ||
NM_016673.2 | NP_057882.2 | |||
AB154246.1 | ||||
AF068615.1 | AAC25711.1 | |||
AK137026.1 | BAE23207.1 | |||
BC046974.1 | AAH46974.1 | |||
BC050928.1 | AAH50928.1 |
More Information
Set Membership: |
Intragenic Non-exonic |