Assay Details
Target Gene Details
Entrez Gene ID: | 14590 |
Gene Name: | gamma-glutamyl hydrolase |
Gene Aliases: |
gamma-GH |
Location: |
Chr.4:20041987-20066111 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ggh | NM_010281.2 | 5 | 539 | NP_034411.2 |
AF051102.1 | 5 | 507 | AAC70002.1 | |
AI846732.1 | 2 | 95 | ||
AK089996.1 | 5 | 496 | ||
BB610405.1 | 5 | 427 | ||
BC009809.1 | 5 | 430 | AAH09809.1 | |
BC150898.1 | 5 | 476 |
More Information
Set Membership: |
Intragenic Exonic |