Assay Details
Target Gene Details
Entrez Gene ID: | 13806 |
Gene Name: | enolase 1, alpha non-neuron |
Gene Aliases: |
0610008I15, AL022784, Eno-1, MBP-1 |
Location: |
Chr.4:150236726-150248873 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eno1 | NM_023119.2 | NP_075608.2 | ||
AK002322.1 | BAC24987.1 | |||
AK002336.1 | BAB22021.1 | |||
AK088788.1 | BAC40572.1 | |||
AK147987.1 | BAE28270.1 | |||
AK150161.1 | BAE29351.1 | |||
AK150192.1 | BAE29369.1 | |||
AK153321.1 | BAE31901.1 | |||
AK177990.1 | ||||
AK186626.1 | ||||
AK190135.1 | ||||
AK190994.1 | ||||
AK191266.1 | ||||
AK194042.1 | ||||
AK194989.1 | ||||
AK195411.1 | ||||
AK197783.1 | ||||
AK200096.1 | ||||
AK202010.1 | ||||
AK202376.1 | ||||
AK202928.1 | ||||
AK203740.1 | ||||
AK204945.1 | ||||
AK207175.1 | ||||
AK207191.1 | ||||
BC003891.1 | AAH03891.1 | |||
BC010685.1 | AAH10685.1 | |||
BC024644.1 | AAH24644.1 | |||
BC039179.1 | ||||
BC056611.1 | AAH56611.1 | |||
BC083334.1 | ||||
BC085098.1 | AAH85098.1 | |||
X52379.1 | CAA36605.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)