Assay Details
Target Gene Details
Entrez Gene ID: | 21985 |
Gene Name: | tumor protein D52 |
Gene Aliases: |
AI043038, mD52 |
Location: |
Chr.3:8928626-9004769 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tpd52 | NM_001025261.1 | NP_001020432.1 | ||
NM_001025262.1 | NP_001020433.1 | |||
NM_001025263.1 | NP_001020434.1 | |||
NM_001025264.1 | NP_001020435.1 | |||
NM_009412.2 | NP_033438.1 | |||
AK005456.1 | BAB24048.1 | |||
AK032111.1 | BAC27709.1 | |||
AK150935.1 | ||||
AK151471.1 | BAE30428.1 | |||
AK151744.1 | BAE30655.1 | |||
AK152543.1 | BAE31298.1 | |||
AK153266.1 | BAE31856.1 | |||
AK153292.1 | BAE31875.1 | |||
AK161753.1 | BAE36558.1 | |||
AK168379.1 | BAE40308.1 | |||
AK168817.1 | BAE40644.1 | |||
AK169751.1 | BAE41345.1 | |||
AK170725.1 | BAE41982.1 | |||
AK171050.1 | BAE42212.1 | |||
AY048852.1 | AAL05266.1 | |||
BC002036.1 | AAH02036.1 | |||
BC004068.1 | AAH04068.1 | |||
BC094018.1 | AAH94018.1 | |||
BE634665.1 | ||||
BG404433.1 | ||||
CA480795.1 | ||||
U44426.1 | AAB40897.1 |