Assay Details
Target Gene Details
Entrez Gene ID: | 75986 |
Gene Name: | agmatine ureohydrolase (agmatinase) |
Gene Aliases: |
5033405N08Rik |
Location: |
Chr.4:141746675-141759263 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Agmat | NM_001081408.1 | 5 | 1004 | NP_001074877.1 |
AK017155.1 | 5 | 901 | ||
BC025916.1 | 2 | 221 | AAH25916.1 | |
BC115693.2 | 5 | 901 |
More Information
Set Membership: |
Intragenic Exonic |