Assay Details
Target Gene Details
Entrez Gene ID: | 58810 |
Gene Name: | aldo-keto reductase family 1, member A1 (aldehyde reductase) |
Gene Aliases: |
2610201A18Rik, Akr1a4 |
Location: |
Chr.4:116636510-116651674 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Akr1a1 | NM_021473.3 | 6 | 948 | NP_067448.1 |
AF225564.1 | 6 | 853 | AAF67111.1 | |
AK005162.1 | 3 | 389 | BAB23853.1 | |
AK005376.1 | 6 | 883 | ||
AK009462.1 | 6 | 725 | BAB26303.1 | |
AK011157.1 | 3 | 389 | BAB27437.1 | |
AK011209.1 | 3 | 389 | BAB27469.1 | |
AK011221.1 | 3 | 388 | ||
AK011321.1 | 6 | 877 | BAB27543.1 | |
AK011388.1 | 6 | 903 | BAB27586.1 | |
AK011667.1 | 6 | 904 | BAB27767.1 | |
AK011794.1 | 6 | 877 | BAB27846.1 | |
AK011856.1 | 6 | 904 | BAB27883.1 | |
AK011906.1 | 6 | 882 | BAB27907.1 | |
AK011908.1 | 6 | 917 | BAB27909.1 | |
AK011918.1 | 6 | 903 | BAB27915.1 | |
AK128906.1 | 6 | 670 | ||
AK144198.1 | 6 | 904 | BAE25762.1 | |
AK146081.1 | 6 | 902 | BAE26885.1 | |
AK146278.1 | 6 | 904 | BAE27036.1 | |
AK152086.1 | 6 | 883 | BAE30936.1 | |
AK159573.1 | 6 | 900 | ||
AK160185.1 | 6 | 878 | ||
AK164461.1 | 6 | 903 | BAE37798.1 | |
AK168950.1 | 6 | 899 | BAE40756.1 | |
AK181349.1 | 2 | 124 | ||
AK184360.1 | 2 | 249 | ||
AK187660.1 | 2 | 261 | ||
AK189553.1 | 2 | 249 | ||
AK196298.1 | 2 | 249 | ||
AK200998.1 | 2 | 236 | ||
AK216201.1 | 2 | 211 | ||
AY207463.1 | 6 | 925 | AAO72145.1 | |
BC039926.1 | 6 | 897 | AAH39926.1 | |
BC046762.1 | 6 | 854 | AAH46762.1 |
More Information
Set Membership: |
Intragenic Exonic |